Learn More
MSC Versaflo Radiant Heat Cover, 3M, Aluminized, 1 EA/Case
Catalog No. 5000684805
Encompass
The 3M Versaflo Radiant Heat Covers are designed for use with 3M Versaflo M-Series helmets to provide protection in high-temperature environments. They shield the head, neck, and shoulders from radiant heat while maintaining the helmet's functionality.
  • Designed for M Series helmets in radiant heat environments
  • Protects the head, neck, and shoulders from radiant heat
  • Made from lightweight aluminized aramid fabric for durability and radiant heat resistance
  • Compatible with flip-up operation of over visor and main visor
  • Modular design supports multiple combinations for tailored coverage levels
  • Suitable for tough and radiant heat applications
Catalog No. 50-006-84805 Supplier MSC Supplier No. 46722062
The 3M Versaflo Radiant Heat Covers are designed for use with 3M Versaflo M-Series helmets to provide protection in high-temperature environments. They shield the head, neck, and shoulders from radiant heat while maintaining the helmet's functionality.
  • Designed for M Series helmets in radiant heat environments
  • Protects the head, neck, and shoulders from radiant heat
  • Made from lightweight aluminized aramid fabric for durability and radiant heat resistance
  • Compatible with flip-up operation of over visor and main visor
  • Modular design supports multiple combinations for tailored coverage levels
  • Suitable for tough and radiant heat applications