Learn More
DuPont™ Tychem™ 10000 Encapsulated Level B
Catalog No. 18888844 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-844 Medium
18-888-845 Small
18-888-843 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-844 Supplier DuPont™ Supplier No. TK527TLYMD000100
Add to Cart
Add to Cart
This item is not returnable. View return policy
10000 Encapsulated Level B offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • 10000 Encapsulated Level B design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Standard 40 mil PVC faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Liquid resistant, extra long zipper to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
TRUSTED_SUSTAINABILITY
Specifications
Color Lime Yellow
Material Tychem
Size Medium
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped