Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 10000 Encapsulated Level B Suit

Catalog No. 18888845 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-845 Small
18-888-844 Medium
18-888-843 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-845 Supplier DuPont™ Supplier No. TK527TLYSM000100
  • $745.00
    $578.65 / Case of 1

    Save $166.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers a high level of protection against toxic, corrosive gases and liquid chemicals

  • 10000 Encapsulated Level B design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes
    • Sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Standard 40 mil PVC face shield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Liquid resistant, extra long zipper to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
TRUSTED_SUSTAINABILITY

Specifications

Color Lime Yellow
For Use With (Application) Industrial, Chemical Handling, Emergency Response
Material Tychem
Size Small
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Level B Suit
Seam Type Taped
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.