Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Complete, fully assembled PAPR Headtop allows for use right out of the box
- Flexible application - Use with any 3M Versaflo system
- Hearing protection compatible with M-985 earmuffs
- Comfortable PAPR alternative to negative pressure respirators and standard faceshields
- Meets the requirements of the ANSI Z87.1-2010 standard for limited eye and face protection devices
- Easy-to-use design with ratchet suspension and liftable visor
- Popular in light industry where respirators and face protection are typically used
- Combine the M-206 with 3M Versaflo PAPR or Supplied Air components to create a complete respiratory assembly.
Specifications
Specifications
| Color | Gray |
| Product Type | Faceshield Assembly |
| For Use With (Application) | Cleaning, Welding, Metal Repair, Grinding, Composite Finishing, Demolition, Painting, Final Finish, Paint Preparation, Assembly and Mechanical, Machine Operations, Sanding |
| For Use With (Equipment) | 3M™ Versaflo™ PAPR and Supplied Air Systems |
| Includes | Comfort Faceseal |
| Model | M-206 |
| Quantity | 1 Ea. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?