Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Respiratory Faceshield Assembly

Catalog No. 19039901 Shop All 3M Products
Change view
Click to view available options
Includes:
Comfort Faceseal
Flame Resistant Faceseal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-039-901 Flame Resistant Faceseal
19-039-900 Comfort Faceseal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-039-901 Supplier 3M™ Supplier No. 7100086969
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Faceshield assembly is a lightweight, comfortable PAPR headtop that combines a respirator and faceshield, with the option of adding earmuffs for hearing protection.

  • Complete, fully assembled PAPR Headtop allows for use right out of the box
  • Flexible application - Use with any 3M Versaflo system
  • Hearing protection compatible with M-985 earmuffs
  • Comfortable PAPR alternative to negative pressure respirators and standard faceshields
  • Meets the requirements of the ANSI Z87.1-2010 standard for limited eye and face protection devices
  • Easy-to-use design with ratchet suspension and liftable visor
  • Popular in light industry where respirators and face protection are typically used
  • Combine the M-206 with 3M Versaflo PAPR or Supplied Air components to create a complete respiratory assembly.
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Product Type Faceshield Assembly
For Use With (Application) Demolition, Grinding, Sanding, Assembly and Mechanical, Cleaning, Machine Operations, Metal Repair
For Use With (Equipment) 3M™ Versaflo™ PAPR and Supplied Air Systems
Includes Flame Resistant Faceseal
Model M-207
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.