Learn More
3M™ Versaflo™ Vortex Cooling Assembly V-100SR with Schrader Fitting
Catalog No. 17018586 Shop All 3M Products
Change view
Click to view available options
Vortex Cooling Assembly V-100SR with Schrader Fitting; Color: Black
Catalog No.
17-018-586 Vortex Cooling Assembly V-100SR with Schrader Fitting; Color: Black
1 options
Catalog No. 17-018-586 Supplier 3M™ Supplier No. 7100066238
V-Series Supplied Air Regulator Valves and Assemblies provide comfortable, cost effective respiratory protection for all-day use.
  • 3M™ Supplied air respirators require a source of clean breathing air such as plant air systems or bottled air
  • Clean, breathable air can be delivered to hoods, helmets, full facepieces, half facepieces and loose-fitting facepieces
  • Supplied air respirators offer many features over traditional respiratory protection, such as cooling or heating incoming air
  • Fit testing only required for tight-fitting facepieces
  • Low maintenance
  • Complete systems include a supplied air hose, an air regulating valve, a breathing tube, and a helmet, hood, or other facepiece
  • Cost-effective way to provide a high level of respiratory protection
  • 3M™'s range of supplied air valves can cool or heat air by up to 50°F (28°C)
  • No filters, cartridges, or batteries to maintain
  • Supplied air provides no additional breathing resistance to the wearer
  • Vortex assembly can cool compressed air by as much as 28°C (50°F)
  • Can operate on pressures ranging from 60 to 90 psig, depending on hose length
  • Includes: Cooling tube with Schrader fittings, Waist belt, Holder; V-100SR with 1/4 in. MPT, 1/4 in. body size, Shrader fitting
  • Product is made to order
  • Lead times for V-100SR assemblies may be longer than other 3M™ Supplied Air Products
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Reusable
Material Plastic
Product Type Cooling Assembly
Size One Size