Learn More
Cytiva Mini capsule VERSAPOR .8/.45um gam 1 pk
Supplier:  Cytiva 12123
Encompass
Use both for prefiltration of particulate liquids prior to final sterilizing filtration, and for removal of particles and debris from liquids not requiring final sterilization. Incorporates two layers of Versapor (acrylic copolymer) membrane: 0.8µm prefilter layer followed by 0.45µm final filter.-Two layers-Barbed hose fittings-Polycarbonate housing This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1093452