Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire VersaHOCI™ Stabilized 0.065% Hypochlorous Acid, Sterile
GREENER_CHOICE

Catalog No. 17010019 Shop All Berkshire Corporation Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
17-010-019 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-010-019 Supplier Berkshire Supplier No. VHOCL6QST
Add to Cart
Add to Cart

VersaHOCl™ Sterile is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire's VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.

VersaHOCl™ Sterile is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire's VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs. Non-toxic with low odor and low residue, VersaHOCl™ establishes the safest standards in disinfectant technology. With an active ingredient of 0.065% hypochlorous acid, it's non-corrosive and effective on hard, non-porous surfaces. This versatile disinfectant is ideal for use in controlled environments such as labs, cleanrooms, compounding pharmacies, and manufacturing facilities and can also be used in areas such as hospitals, clinics, healthcare facilities, schools, and government and office buildings.

  • Kills common microbiological contaminations on surfaces
  • Very low residue, saving time on residue removal
  • Wide range of material compatibility
  • Wide range of material compatibility
  • Wide range of material compatibility
  • No special disposal is required
  • Bottles individually bagged
  • Facilitates transfer disinfection into cleanroom
  • Can be used safely on all hard, non-porous surfaces
  • Ready to Use - No mixing required
  • Not classed as corrosive

Specifications

Product Type Cleaner/Disinfectant
Form Clear Liquid
pH Neutral
Sterility Sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Clear
Composition 0.065% Hypochlorous Acid
Disposable Single-use
Filtered Non-filtered
Flammable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.