Learn More
Berkshire VersaHOCI™ Stabilized 0.065% Hypochlorous Acid
GREENER_CHOICE
Catalog No. 17018061 Shop All Berkshire Corporation Products
Change view
Click to view available options
Scent:
Unscented
Catalog No. Scent
17-018-061 Unscented
1 options
This item is not returnable. View return policy
Catalog No. 17-018-061 Supplier Berkshire Supplier No. VHOCL6Q
Add to Cart
Add to Cart
This item is not returnable. View return policy
VersaHOCl™ is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
  • Kills common microbiological contaminations on surfaces
  • Very low residue, saving time on residue removal
  • Wide range of material compatibility
  • No special disposal is required
  • Bottles individually bagged
  • Facilitates transfer disinfection into cleanroom
  • Can be used safely on all hard, non-porous surfaces
  • Ready to Use
  • No mixing required
  • Not classed as corrosive
Specifications
Product Type Cleaner and Disinfectant Solution
Form Liquid Spray
Handle Material Polypropylene
Handle Type Trigger
Product Line VersaHOCI
Tip Type Spray/Stream/Off Nozzle
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color White
Container Type Bottle
Disposable Reusable
Filtered Non-filtered
Flammable No
Packaging Individually Bagged