Learn More
Thermo Scientific™ VersaLid for VersaCool™ Refrigerated Circulating Bath, reversible hinged lid
Catalog No. 13275107 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cover
Catalog No. Type
13-275-107 Cover
1 options
Catalog No. 13-275-107 Supplier Thermo Scientific™ Supplier No. 230000002
Reversible hinged lid to be used with Thermo Scientific VersaCool Refrigerated Circulating Bath.
Utilize the VersaLid for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Configurable to the user’s comfort, the VersaLid offers a tool-less setting for left or right-handed hinge movement and also for no hinge movement. It’s dripless when used in hinge mode.
  • One lid that can be left-handed hinge, right-handed hinge, or unhinged lid 
  • Drip-less lid reduces moisture and directs condensate back into bath
TRUSTED_SUSTAINABILITY
Specifications
Description VersaLid-reversible hinged lid
For Use With (Equipment) Thermo Scientific VersaCool Refrigerated Circulating Bath
Type Cover