Learn More
Mitegen VersaPins All-In-One Tool for Home Source Set-up
SDP
Catalog No. 501941076
Encompass_Preferred
The patented VersaPin for home source allows you to perform several operations needed to prepare your home source for diffraction measurements. The VersaPin performs the role of several tools and will save time while improving the accuracy of your measurements. Applications for the Versapin include: align the sample rotation axis; visualize the X-ray beam; precisely determine beam coordinates; align the sample in the beam; center the beamstop on the beam; calibrate the sample-to-detector distance; calibrate the monochromator energy. The VersaPin is manufactured and sold by MiTeGen under exclusive license from Argonne National Laboratory.
Catalog No. 501941076 Supplier Mitegen Supplier No. VERSAPINHS01
The patented VersaPin for home source allows you to perform several operations needed to prepare your home source for diffraction measurements. The VersaPin performs the role of several tools and will save time while improving the accuracy of your measurements. Applications for the Versapin include: align the sample rotation axis; visualize the X-ray beam; precisely determine beam coordinates; align the sample in the beam; center the beamstop on the beam; calibrate the sample-to-detector distance; calibrate the monochromator energy. The VersaPin is manufactured and sold by MiTeGen under exclusive license from Argonne National Laboratory.