Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire VersaQuat™ RTU One-Step Disinfectant and Cleaner

Catalog No. 17010018 Shop All Berkshire Corporation Products
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
17-010-018 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-010-018 Supplier Berkshire Supplier No. VQUAT4G
Add to Cart
Add to Cart

VersaQuat™ delivers consistent quality and intuitive usage. Recommended for use as a standard disinfectant as part of a disinfectant rotation program.

VersaQuat™ is an EPA-registered, RTU (ready-to-use) one-step disinfectant and cleaner which utilizes well-established and accepted quaternary ammonium chemistry with acceptable residue and safety profiles. With broad-spectrum, best-in-class efficacy claims (30 kill claims), VersaQuat™ delivers consistent quality and intuitive usage. Recommended for use as a standard disinfectant as part of a disinfectant rotation program, VersaQuat™ is ideal for use in laboratories, hospitals, home healthcare settings, and veterinary clinics and can also be used in offices.

  • Kills common microbiological contaminations on surfaces
  • Can use in a variety of application
  • Ideal for large surface cleaning
  • No strong odor provides good user acceptability
  • No mixing required
  • No special disposal is required

Specifications

Product Type Cleaner/Disinfectant
Form Clear Liquid
Handle Material Polypropylene
Handle Type Trigger
pH Neutral
Tip Type Spray, Stream, Off Nozzle
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Clear
Composition 0.75% C9H14ClN, 0.75% C23H42ClN
Disposable Single-use
Filtered Non-filtered
Flammable No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.