Learn More
Thermo Scientific™ VersaRack for VersaCool™ Refrigerated Circulating Bath, Stainless steel, adjustable
GSA_VA
Catalog No. 13275101 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Tube Rack
Catalog No. Type
13-275-101 Tube Rack
1 options
Catalog No. 13-275-101 Supplier Thermo Scientific™ Supplier No. 230000001
Stainless steel adjustable rack. For use with Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Utilize the VersaRack for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
With the ability to accommodate a wide range of beaker and test tube sizes, the VersaRack offers a tool-less adjustable height setting . Its drip-less features allows it to rests over the bath opening saving you valuable bench space.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description VersaRack - Stainless steel adjustable rack
For Use With (Equipment) Thermo Scientific VersaCool Refrigerated Circulating Bath
Type Tube Rack
Holds Holds up to 75 test Tubes that are 10 mm dia.
Holds up to 48 test Tubes that are 16 mm dia.
Holds up to 21 test Tubes that are 25 mm dia.