Learn More
MSC Clorox Healthcare Wipes, Alcohol-Free, 12 x 12 in, 110 Count
Catalog No. 502836543
Encompass
Clorox Healthcare VersaSure Cleaner Disinfectant Wipes are alcohol-free, 1-ply wipes measuring 12 x 12 inches, with 110 wipes per bucket. These ready-to-use wipes are EPA registered to kill over 40 microorganisms, including antibiotic-resistant organisms, in as little as 30 seconds, making them ideal for commercial and healthcare settings.
  • EPA registered to kill more than 40 microorganisms, including 11 antibiotic resistant organisms.
  • Noncorrosive and contain no added fragrances, chemical odors, fumes or bleach.
  • Disinfects most bacteria and viruses in as little as 30 seconds.
  • Ready-to-use one-step cleaner disinfectant wipes.
  • Intended for use in commercial settings.
  • Alcohol-free wipes.
  • Designed for patient and staff comfort.
Catalog No. 50-283-6543 Supplier MSC Supplier No. 65372906
Clorox Healthcare VersaSure Cleaner Disinfectant Wipes are alcohol-free, 1-ply wipes measuring 12 x 12 inches, with 110 wipes per bucket. These ready-to-use wipes are EPA registered to kill over 40 microorganisms, including antibiotic-resistant organisms, in as little as 30 seconds, making them ideal for commercial and healthcare settings.
  • EPA registered to kill more than 40 microorganisms, including 11 antibiotic resistant organisms.
  • Noncorrosive and contain no added fragrances, chemical odors, fumes or bleach.
  • Disinfects most bacteria and viruses in as little as 30 seconds.
  • Ready-to-use one-step cleaner disinfectant wipes.
  • Intended for use in commercial settings.
  • Alcohol-free wipes.
  • Designed for patient and staff comfort.