Learn More
MSC Clorox Healthcare VersaSure Cleaner Disinfectant Wipes, Alcohol-Free, 85 Count, 6.75 x 8 in
Catalog No. 502839457
Encompass
Clorox Healthcare VersaSure Cleaner Disinfectant Wipes are alcohol-free quat wipes designed for broad-spectrum disinfection in various healthcare settings. They kill over 40 pathogens, including MRSA and TB, in 2 minutes or less, and feature a low-odor, low-residue formula compatible with common healthcare surfaces.
  • Alcohol-free quat wipes
  • Versatile for use in various settings
  • Provides broad-spectrum disinfection
  • Kills bacteria, viruses, and fungi in 2 minutes or less
  • Low residue formula
  • One-step disinfectant wipes
  • Kills over 40 pathogens in 2 minutes or less
  • Patented technology enhances quat activity
  • Low-odor
  • Designed for compatibility with common healthcare surfaces
  • 85 count canister
  • Wipe size: 6.75 x 8 in
Catalog No. 50-283-9457 Supplier MSC Supplier No. 65354367
Clorox Healthcare VersaSure Cleaner Disinfectant Wipes are alcohol-free quat wipes designed for broad-spectrum disinfection in various healthcare settings. They kill over 40 pathogens, including MRSA and TB, in 2 minutes or less, and feature a low-odor, low-residue formula compatible with common healthcare surfaces.
  • Alcohol-free quat wipes
  • Versatile for use in various settings
  • Provides broad-spectrum disinfection
  • Kills bacteria, viruses, and fungi in 2 minutes or less
  • Low residue formula
  • One-step disinfectant wipes
  • Kills over 40 pathogens in 2 minutes or less
  • Patented technology enhances quat activity
  • Low-odor
  • Designed for compatibility with common healthcare surfaces
  • 85 count canister
  • Wipe size: 6.75 x 8 in