Learn More
MSC Clorox Healthcare VersaSure Disinfectant Wipes, Alcohol-Free, 1-Ply, 12 x 10 in
Catalog No. 502836542
Encompass
These disinfectant wipes provide an alcohol-free solution for cleaning and disinfecting a broad range of hard surfaces. The durable, cloth-like wipes are EPA registered to kill over 40 pathogens, including bacteria and viruses, in as fast as 30 seconds. Designed for healthcare settings, they offer one-step disinfection with no harsh chemical odors or added fragrances, ensuring patient and staff comfort.
  • Kills over 40 pathogens, including bacteria and viruses.
  • EPA registered for hospital disinfection.
  • Alcohol-free and bleach-free formula.
  • No harsh chemical odors, fumes, or added fragrances.
  • Nonwoven wipe material does not leave lint.
  • Strong and durable for effective cleaning.
  • Good compatibility with common healthcare surfaces.
Catalog No. 50-283-6542 Supplier MSC Supplier No. 65372872
These disinfectant wipes provide an alcohol-free solution for cleaning and disinfecting a broad range of hard surfaces. The durable, cloth-like wipes are EPA registered to kill over 40 pathogens, including bacteria and viruses, in as fast as 30 seconds. Designed for healthcare settings, they offer one-step disinfection with no harsh chemical odors or added fragrances, ensuring patient and staff comfort.
  • Kills over 40 pathogens, including bacteria and viruses.
  • EPA registered for hospital disinfection.
  • Alcohol-free and bleach-free formula.
  • No harsh chemical odors, fumes, or added fragrances.
  • Nonwoven wipe material does not leave lint.
  • Strong and durable for effective cleaning.
  • Good compatibility with common healthcare surfaces.