Learn More
Anton Paar Versatile model for R&D and high-end QC applicationsTorque range from 0.2 nNm to 230 mNmCompatible with 200+ plug-and-play accessoriesSupports rheology, tribology, powder, DMA, and UTMSuccessor to MCR 302e
Catalog No. 5000580515
Encompass_Preferred
MCR 503 SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: 360° capacitive sensor, non-contacting, fully integrated in bearingWorking Modules: CMTMin. torque (rotation): 1 nNmMin. torque (oscillation): 0.2 nNmMax. torque: 230 mNmTorque resolution: 0.05 nNmAngular deflection resolution: < 1 nradMin. angular velocity: 0 rad/sMax. angular velocity / max. speed: 314 rad/s (3,000 rpm)Min. frequency: 2 x 10-8 HzMax. frequency: 200 HzNormal force range: 0.001 N to 50 NNormal force resolution: 0.1 mNTruStrain: YesReady for DMA in tension, bending, and compression: Yes
Catalog No. 50-005-80515 Supplier Anton Paar Supplier No. 220278
Please contact your sales representative for a quote or further ordering instructions.
MCR 503 SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: 360° capacitive sensor, non-contacting, fully integrated in bearingWorking Modules: CMTMin. torque (rotation): 1 nNmMin. torque (oscillation): 0.2 nNmMax. torque: 230 mNmTorque resolution: 0.05 nNmAngular deflection resolution: < 1 nradMin. angular velocity: 0 rad/sMax. angular velocity / max. speed: 314 rad/s (3,000 rpm)Min. frequency: 2 x 10-8 HzMax. frequency: 200 HzNormal force range: 0.001 N to 50 NNormal force resolution: 0.1 mNTruStrain: YesReady for DMA in tension, bending, and compression: Yes