Learn More
Anton Paar Versatility that meets your needs
Catalog No. 5000580503
Encompass_Preferred
Modular cell concept for any type of sampleCompatibility with a range of ATR and transmission cellsAutomatic cell module exchange recognitionIdentification and verification of any sample or substanceCustom library creation in just a few clicksVariety of industry standard libraries to choose fromQuantification method creation in under three minutes*
Catalog No. 50-005-80503 Supplier Anton Paar Supplier No. 387366
Modular cell concept for any type of sampleCompatibility with a range of ATR and transmission cellsAutomatic cell module exchange recognitionIdentification and verification of any sample or substanceCustom library creation in just a few clicksVariety of industry standard libraries to choose fromQuantification method creation in under three minutes*