Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersaTREK™ Myco Antibiotic Supplement (Myco AS)
GSA_VA

Catalog No. Y711442 Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging Type:
25 mL Each
Media Type:
Myco antibiotic supplement
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Media Type Quantity
Y711442 25 mL Each Myco antibiotic supplement 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. Y711442 Supplier Thermo Scientific™ Supplier No. Y711442
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accurately detect mycobacteria in potentially contaminated specimens using a five-antibiotic formula: polymyxin B, azlocillin, fosfomycin, nalidixic acid and amphotericin B.

Accurately detect mycobacteria in potentially contaminated specimens using the Thermo Scientific™ VersaTREK™ Myco Antibiotic Supplement (Myco AS). VersaTREK Myco AS includes five antibiotics: polymyxin B, azlocillin, fosfomycin, nalidixic acid and amphotericin B.

The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.

Not all products are available for sale in all territories. Please inquire.

VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description VersaTREK Myco Antibiotic Supplement (Myco AS)
Media Type Myco antibiotic supplement
Product Type Microbiology Media
Target Organism Class Mycobacteria
Test Type Microbial Detection, Antimicrobial Susceptibility
No. per Pack 5
Packaging Type 25 mL Each
Quantity 25 mL
Specimen Type All Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
Species Mycobacterium tuberculosis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.