Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersaTREK™ Myco Media
GSA_VA

Catalog No. y711142
Change view
Click to view available options
Product Type:
Microbiology Media
Quantity:
11.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
Y711142 Microbiology Media 11.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. Y711142 Supplier Thermo Scientific™ Supplier No. Y711142
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reliably detect a wide variety of mycobacteria species from all specimen types with one bottle using a unique growth matrix, cellulose sponges, to provide better recovery.

Reliably detect a wide variety of mycobacteria species from all specimen types with just one bottle using Thermo Scientific™ VersaTREK™ Myco Media. VersaTREK Myco Media utilizes a unique growth matrix, via cellulose sponges, to provide better recovery of mycobacteria species while the VersaTREK instrument's detection technology automatically incubates and continuously monitors inoculated bottles for reliable, automated myco testing.

The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.

Mycobacteria detection also requires VersaTREK Myco Growth Supplement (Myco GS, Part No. 7112-42) and VersaTREK Myco Antibiotic Supplement (Myco AS, Part No. 7114-42).

Not all products are available for sale in all territories. Please inquire.

VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store away from direct sunlight
Description VersaTREK Myco Media
Media Type Modified Middlebrook 7H9 Broth with Sponges
Product Type Microbiology Media
Target Organism Class Mycobacteria
Test Type Microbial Detection, Antimicrobial Susceptibility
No. per Pack 50
Packaging Type 11.5 mL Each
Quantity 11.5 mL
Specimen Type Blood, Sterile Body Fluid, Processed Specimens
Volume (Metric) Specimen Up to 1 mL
Species Mycobacterium tuberculosis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.