Learn More
Description
Perform accurate, FDA-cleared Mtb susceptibility testing utilizing pyrazinamide (PZA) with the Thermo Scientific™ VersaTREK™ Myco PZA Kit.
The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | VersaTREK Myco PZA Kit |
| Product Type | Microbiology Media |
| Target Organism Class | Mycobacteria |
| Test Type | Microbial Detection, Antimicrobial Susceptibility |
| Packaging Type | 50 Tests/Kit |
| Quantity | 2 x 25 mL Pyrazinamide, 4 x 25 mL PZA Rehydration Buffer |
| Specimen Type | All Samples (e.g. Sputum, Blood, Tissue, Urine, etc.) |
| Species | Mycobacterium tuberculosis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.