Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersaTREK™ REDOX™ Media

Catalog No. Y710244 Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging Type:
80 mL/Bottle
Media Type:
VersaTREK REDOX 1 Media
VersaTREK REDOX 2 Media
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Media Type
Y710244 80 mL/Bottle VersaTREK REDOX 1 Media
Y710344 80 mL/Bottle VersaTREK REDOX 2 Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. Y710244 Supplier Thermo Scientific™ Supplier No. Y710244
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recover organisms from all patient scenarios, including pediatrics, with just two bottles, reducing inventory and costs and enhancing fastidious organism detection.

Just two bottles are all you need to recover organisms from all patient scenarios, reducing inventory and costs, with Thermo Scientific™ VersaTREK™ REDOX™ Media. There's no need for costly specialty media, as REDOX 1 aerobic media and REDOX 2 anaerobic media use dilution and timing to detect microbes and are the only media FDA-cleared for draws as low as 0.1mL, perfect for pediatric patients. Plus, the highly-enriched REDOX media supports a wide range of organisms, enhancing the VersaTREK System's ability to better recover fastidious organisms.

Meet the demands of both high and low volume laboratories with the Thermo Scientific VersaTREK System, which comes with a full line of ancillary and accessory products. With four FDA-cleared tests and a simple, two-bottle media system, the VersaTREK System provides maximum value with faster results and fewer limitations. 
  • Largest blood-to-broth dilution ratio in the industry (1:9) to dilute out the effects of antimicrobials, serum host factors and other agents
  • Enhanced, enriched media to support the wide range of organisms found in the microbiology laboratory
  • Available in 40 and 80mL sizes and suitable for all patient populations
  • Only system capable of detecting any gas produced or consumed by organisms, allowing detection of a wider range of both common and fastidious organisms
  • Simplified workflow includes easy scan function for bottle entry, intuitive icons to reduce training time, and no daily QC requirement for maximum efficiency

Not all products are available for sale in all territories. Please inquire. 

VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store away from direct sunlight
Media Type VersaTREK REDOX 1 Media
Product Type Blood Culture Medium
Target Organism Class Aerobic Microorganisms
Test Type Microbial Detection
Manual/Automated Automated
No. per Pack 50 Bottles/Box
Packaging Type 80 mL/Bottle
Quantity 80 mL
Specimen Type Blood, Sterile Body Fluid
Volume (Metric) Specimen 0.1 to 10 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.