Learn More
StatLab Versenate Decal Solution EDTA
SDP
Catalog No. 21033360
Change view
Click to view available options
Solution Type:
Decalcifier
Catalog No. Solution Type
21-033-360 Decalcifier
1 options
Catalog No. 21-033-360 Supplier StatLab Supplier No. DCVERGAL
Add to Cart
Add to Cart
A non-aggressive chelating bone decalcifier, allows extensive use of special stains and immuno-histochemical techniques
  • A non-aggressive chelating bone decalcifier which allows extensive use of special stains and immuno-histochemical techniques
  • Excellent when iron stains are required
  • This decal solution can take several days to complete the decalcification process
Specifications
Product Type Decalcifying Solution
Solution Type Decalcifier
Form Solution