Learn More
StatLab Versenate Decal Solution
SDP
Catalog No. 22000034
Change view
Click to view available options
Solution Type:
Decalcifier
Catalog No. Solution Type
22-000-034 Decalcifier
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-034 Supplier StatLab Supplier No. DCVERLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
A non-aggressive chelating bone decalcifier which allows extensive use of special stains and immuno-histochemical techniques
  • Excellent when iron stains are required
  • This decal solution can take several days to complete the decalcification process
Specifications
Product Type Decalcifying Solution
Solution Type Decalcifier
Form Solution