Learn More
Analytical Sales & Services Versi-Rac
Catalog No. 5000806222
Encompass
96-Well Versi-Rac, Assorted Colors This rack accommodates 96 0.5mL microcentrifuge tubes on one side, or 96 1.5-2.0mL microcentrifuge tubes on the other. The rack is arranged in the standard 8x12 array. It features a handy snap-in-place lid, and easy to grasp tabs on each end of the rack. Grid lines are molded on the side of the rack that accommodates the smaller tubes making it easier to see the wells. Raised rings are molded on the other side of the rack making it easier to replace the 1.5-2.0mL tubes from those wells.
Catalog No. 50-008-06222 Supplier Analytical Sales & Services Supplier No. 96152
96-Well Versi-Rac, Assorted Colors This rack accommodates 96 0.5mL microcentrifuge tubes on one side, or 96 1.5-2.0mL microcentrifuge tubes on the other. The rack is arranged in the standard 8x12 array. It features a handy snap-in-place lid, and easy to grasp tabs on each end of the rack. Grid lines are molded on the side of the rack that accommodates the smaller tubes making it easier to see the wells. Raised rings are molded on the other side of the rack making it easier to replace the 1.5-2.0mL tubes from those wells.