Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Versican isoform V0 protein, human (recombinant, His-B2M-Myc tagged) | >95.0% | 100UG
SDP

Catalog No. 5000175817
Encompass_Preferred

Versican isoform V0 protein is a recombinant, partially truncated human extracellular matrix proteoglycan supplied as a lyophilized powder with N-terminal 10xHis and B2M tags and a C-terminal Myc tag. The preparation is endotoxin-controlled and >95% pure, intended for research into hyaluronan binding, fibulin-1 interactions, and cell-matrix signaling and motility.

  • Recombinant human versican V0 partial construct.
  • Lyophilized powder formulated with trehalose for stability.
  • N-terminal 10xHis and B2M tags, C-terminal Myc tag.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg as determined by LAL.
  • Recommended storage at -20°C; aliquot and freeze at -80°C for long term.
  • Suitable for studies of extracellular matrix binding and cell motility.

Catalog No. 50-001-75817 Supplier Medchemexpress LLC Supplier No. HYP702616100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Versican isoform V0 protein is a recombinant, partially truncated human extracellular matrix proteoglycan supplied as a lyophilized powder with N-terminal 10xHis and B2M tags and a C-terminal Myc tag. The preparation is endotoxin-controlled and >95% pure, intended for research into hyaluronan binding, fibulin-1 interactions, and cell-matrix signaling and motility.

  • Recombinant human versican V0 partial construct.
  • Lyophilized powder formulated with trehalose for stability.
  • N-terminal 10xHis and B2M tags, C-terminal Myc tag.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <1 EU/μg as determined by LAL.
  • Recommended storage at -20°C; aliquot and freeze at -80°C for long term.
  • Suitable for studies of extracellular matrix binding and cell motility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.