Learn More
Research Products International Corp Versi-Dry Lab Soakers, Standard Absorbant, White, 18 x 20 Inch Mats, 50 Mats per Box
SDP
Catalog No. 50212644
Encompass
Versi-Dry Lab Soakers offer the best value and protection for covering all work surfaces. Absorb spills quickly and provides a soft thick cushion for fragile items. Standard Versi-Dry absorbs up to 750 ml/m2.
Features a highly durable blue top layer that remains intact even when wet. Mats have a skid resistant polyethylene backing.
Catalog No. 50-212-644 Supplier Research Products International Corp Supplier No. 248896
Versi-Dry Lab Soakers offer the best value and protection for covering all work surfaces. Absorb spills quickly and provides a soft thick cushion for fragile items. Standard Versi-Dry absorbs up to 750 ml/m2.
Features a highly durable blue top layer that remains intact even when wet. Mats have a skid resistant polyethylene backing.