Learn More
MSC Betco All-Purpose Cleaner, 2 L, Bottle, Disinfectant, Fresh Scent
Catalog No. 502837154
Encompass
This hospital grade disinfectant kills a broad spectrum of bacteria and viruses. The power of peroxide makes this an environmentally friendly, all-purpose solution that is effective and safe to use on most non-porous surfaces. It is a resourceful multi-purpose solution for cleaning, disinfecting, and deodorizing, providing clinical level results with a neutral pH at dilution.
  • Hospital grade disinfectant.
  • Kills a broad spectrum of bacteria and viruses, including SARS-CoV-2, HIV-1, and Influenza A.
  • Contains hydrogen peroxide, making it environmentally friendly.
  • Effective and safe to use on most non-porous surfaces.
  • Multi-purpose for cleaning, disinfecting, and deodorizing.
  • Neutral pH at dilution.
  • Saves time, labor, and cost.
Catalog No. 50-283-7154 Supplier MSC Supplier No. 72529555
This hospital grade disinfectant kills a broad spectrum of bacteria and viruses. The power of peroxide makes this an environmentally friendly, all-purpose solution that is effective and safe to use on most non-porous surfaces. It is a resourceful multi-purpose solution for cleaning, disinfecting, and deodorizing, providing clinical level results with a neutral pH at dilution.
  • Hospital grade disinfectant.
  • Kills a broad spectrum of bacteria and viruses, including SARS-CoV-2, HIV-1, and Influenza A.
  • Contains hydrogen peroxide, making it environmentally friendly.
  • Effective and safe to use on most non-porous surfaces.
  • Multi-purpose for cleaning, disinfecting, and deodorizing.
  • Neutral pH at dilution.
  • Saves time, labor, and cost.