Learn More
Accuflow Systems Saint-Gobain Versilon Tubing, Ether-Based Polyurethane, 3/8 in ID, 100 ft
Catalog No. NC2470379
Encompass_Preferred
Braid-reinforced ether-based polyurethane tubing designed for elevated working pressures and high abrasion environments. The tubing remains flexible at extreme low temperatures and is suitable for large-volume transfer of viscous fluids, pastes, and slurries.
  • Ether-based polyurethane material.
  • 3/8 in inside diameter, 5/8 in outside diameter, 1/8 in wall thickness.
  • Braid reinforcement for elevated working pressure.
  • 100 ft package length.
  • Clear color.
  • Hardness 85 Shore A (15 sec).
  • Max working pressure 205 psi at 73°F, 115 psi at 180°F.
  • Retains flexibility to -100°F (-73°C).
Catalog No. NC2470379 Supplier Accuflow Systems Supplier No. AZY02029
Braid-reinforced ether-based polyurethane tubing designed for elevated working pressures and high abrasion environments. The tubing remains flexible at extreme low temperatures and is suitable for large-volume transfer of viscous fluids, pastes, and slurries.
  • Ether-based polyurethane material.
  • 3/8 in inside diameter, 5/8 in outside diameter, 1/8 in wall thickness.
  • Braid reinforcement for elevated working pressure.
  • 100 ft package length.
  • Clear color.
  • Hardness 85 Shore A (15 sec).
  • Max working pressure 205 psi at 73°F, 115 psi at 180°F.
  • Retains flexibility to -100°F (-73°C).