Learn More
Accuflow Systems Versilon Tubing, Thermoplastic Elastomer, 1/4 in ID, 3/8 in OD
Supplier:  Accuflow Systems AJD00017
Encompass_Preferred
Clear, flexible thermoplastic elastomer tubing with an FEP inner liner for inert chemical transfer and general fluid handling. It provides chemical resistance, good bend radius, and visual clarity for monitoring flow.
  • 1/4 in inside diameter, 3/8 in outside diameter, 1/16 in wall thickness.
  • 50 ft coil length.
  • Thermoplastic elastomer construction with FEP inner liner for chemical inertness.
  • 66 Shore A hardness for flexibility and kink resistance.
  • Maximum temperature 165 °F (74 °C); working pressure approximately 55 psi at 73 °F.
  • Crystal clarity for visual flow inspection.
  • Common applications: chemical processing, solvent and adhesive transfer, food and beverage dispensing, laboratory fluid handling.
Catalog No. 50-236-5813