Learn More
Accuflow Systems Saint-Gobain Versilon Tubing, SE-200, FEP Liner, 3/16 in ID
Catalog No. 502365795
Encompass_Preferred
Versilon SE-200 tubing is an FEP-lined, thermoplastic elastomer flexible tubing designed for chemical transfer and fluid handling. The FEP inner liner provides inertness and broad chemical compatibility while the elastomer outer delivers flexibility and an outstanding bend radius. This configuration is 3/16 in (4.76 mm) ID x 5/16 in (7.94 mm) OD, 0.063 in (1.59 mm) wall, and 50 ft length.
  • FEP inner liner provides broad chemical compatibility and inertness.
  • Thermoplastic elastomer outer for flexibility and resistance to kinking.
  • 3/16 in ID, 5/16 in OD, 0.063 in wall, 50 ft length.
  • Crystal clear for visual monitoring of fluids.
  • Phthalate-free and free of BPA.
  • Suitable for chemical processing, solvent and adhesive transfer, and semiconductor applications.
Catalog No. 50-236-5795 Supplier Accuflow Systems Supplier No. AJD00012
Versilon SE-200 tubing is an FEP-lined, thermoplastic elastomer flexible tubing designed for chemical transfer and fluid handling. The FEP inner liner provides inertness and broad chemical compatibility while the elastomer outer delivers flexibility and an outstanding bend radius. This configuration is 3/16 in (4.76 mm) ID x 5/16 in (7.94 mm) OD, 0.063 in (1.59 mm) wall, and 50 ft length.
  • FEP inner liner provides broad chemical compatibility and inertness.
  • Thermoplastic elastomer outer for flexibility and resistance to kinking.
  • 3/16 in ID, 5/16 in OD, 0.063 in wall, 50 ft length.
  • Crystal clear for visual monitoring of fluids.
  • Phthalate-free and free of BPA.
  • Suitable for chemical processing, solvent and adhesive transfer, and semiconductor applications.