Learn More
GeminiBio™ GemCell™ Normal Human Serum
SDP
Catalog No. 11852133 Shop All Gemini Bio Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
11-852-133 100 mL
1 options
Catalog No. 11-852-133 Supplier GeminiBio™ Supplier No. 100110100
Add to Cart
Add to Cart
GemCell™ Normal Human Serum is a defibrinated, sterile-filtered human serum from healthy U.S. donors of all ABO blood types, collected at FDA-licensed facilities. Tested non-reactive for viral markers, it offers consistent, low-bioburden performance for cell culture and research.
GemCell™ Normal Human Serum is a defibrinated, sterile-filtered human serum from healthy U.S. donors of all ABO blood types, collected at FDA-licensed facilities. Tested non-reactive for viral markers, it offers consistent, low-bioburden performance for cell culture and research.
Specifications
Form Serum
Format Bottle
Product Type Human Serum
Quantity 100 mL
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Cell Culture, further Manufacturing, or Research use only. Not for direct Therapeutic use.