Learn More
MSC Simpson Axial Cam Pump Kit, 3300 PSI, 2.4 GPM, Vertical
Catalog No. 502837620
Encompass
This axial cam pump kit is designed for optimal performance, offering high efficiency while minimizing engine load. It features stainless steel pistons for enhanced seal life and is equipped with high-performance double high-pressure seals. The maintenance-free crankcase is manufactured from fully anodized die-cast aluminum, providing protection against corrosion and sealing against contamination.
  • Comes fully plumbed
  • Threaded M22 male connection
  • Double seal system prevents water leaks
  • Thermal relief protection to prevent the pump from overheating
  • Gas engine flange fits most engines
  • Designed to fit most makes and models of cold water gas pressure washers with horizontal engines
  • Includes a 1-year limited warranty
Catalog No. 50-283-7620 Supplier MSC Supplier No. 49039688
This axial cam pump kit is designed for optimal performance, offering high efficiency while minimizing engine load. It features stainless steel pistons for enhanced seal life and is equipped with high-performance double high-pressure seals. The maintenance-free crankcase is manufactured from fully anodized die-cast aluminum, providing protection against corrosion and sealing against contamination.
  • Comes fully plumbed
  • Threaded M22 male connection
  • Double seal system prevents water leaks
  • Thermal relief protection to prevent the pump from overheating
  • Gas engine flange fits most engines
  • Designed to fit most makes and models of cold water gas pressure washers with horizontal engines
  • Includes a 1-year limited warranty