Learn More
Air Science Usa Vertical Clean Bench 48" Wide with UV System
SDP
Catalog No. NC2684335
Encompass_Preferred
Vertical Laminar Flow Cabinet 48" Wide With UV Lamp and front cover. Protects your non-hazardous applications with Vertical unidirectional (laminar) ISO Class 4 air across the work area. Includes Stainless Steel Workurface. Cart-Bw-Vlf-48 Base Stand avilable and sold separatly
Catalog No. NC2684335 Supplier Air Science Usa Supplier No. VLF48A(UV)
Vertical Laminar Flow Cabinet 48" Wide With UV Lamp and front cover. Protects your non-hazardous applications with Vertical unidirectional (laminar) ISO Class 4 air across the work area. Includes Stainless Steel Workurface. Cart-Bw-Vlf-48 Base Stand avilable and sold separatly