Learn More
USA Scientific Inc Vert High Speed Tube Holder
Catalog No. 502745397
Encompass
Vertical high speed tube holder for Vortex-Genie 2, holds nine 1.5 or 2.0 mL tubes
Catalog No. 50-274-5397 Supplier USA Scientific Inc Supplier No. 74008209
Vertical high speed tube holder for Vortex-Genie 2, holds nine 1.5 or 2.0 mL tubes