Learn More
Bellco Glass, Inc. Vertical Sidearm Sample System, 45mm
SDP
Catalog No. 501944737
Encompass_Preferred
Vertical Sidearm Sample System,45mm. The Bellco sample system attaches to gl45 nozzle allowing the user to take repeated samples into a standard 50ml disposable centrifuge tube. The sampler is designed to connect to an existing pipe by mounting under the open cap. Samples are taken by evacuating air with the supplied syringe & filter, thereby drawing culture into the 50ml disposable cent. tube. The Bellco Sampler can be used on third party vessels that meet the arm and clearance dimensions. MADE IN THE USA
Catalog No. 50-194-4737 Supplier Bellco Glass, Inc. Supplier No. 196403000
Vertical Sidearm Sample System,45mm. The Bellco sample system attaches to gl45 nozzle allowing the user to take repeated samples into a standard 50ml disposable centrifuge tube. The sampler is designed to connect to an existing pipe by mounting under the open cap. Samples are taken by evacuating air with the supplied syringe & filter, thereby drawing culture into the 50ml disposable cent. tube. The Bellco Sampler can be used on third party vessels that meet the arm and clearance dimensions. MADE IN THE USA