Learn More
MSC SPI Vertical Dial Test Indicator, 0.0300 in Max, 1 ea
Catalog No. 501920080
Encompass
This Vertical Dial Test Indicator is designed for precise measurement of geometric tolerances and dimensional deviations in workpieces, particularly in confined spaces. It offers universal positioning and comes with a short form calibration certificate.
  • Easy-to-read dial
  • Universal positioning capability
  • Jeweled bearing movement
  • Supplied with 3/8 in dovetail stems
  • Includes a short form calibration certificate
  • Furnished in a fitted case
  • Two year limited warranty
  • Measures up to 0.0300 in
  • Accuracy of 0.000500 in
  • Includes 2 contact points
Catalog No. 50-192-0080 Supplier MSC Supplier No. 37543931
This Vertical Dial Test Indicator is designed for precise measurement of geometric tolerances and dimensional deviations in workpieces, particularly in confined spaces. It offers universal positioning and comes with a short form calibration certificate.
  • Easy-to-read dial
  • Universal positioning capability
  • Jeweled bearing movement
  • Supplied with 3/8 in dovetail stems
  • Includes a short form calibration certificate
  • Furnished in a fitted case
  • Two year limited warranty
  • Measures up to 0.0300 in
  • Accuracy of 0.000500 in
  • Includes 2 contact points