Learn More
Selleck Chemical LLC Verteporfin 10mg 129497-78-5 CL 318952, Visudyne
SDP
Supplier:  Selleck Chemical LLC S178610MG
Encompass_Preferred
Verteporfin (CL 318952, Visudyne) is a small molecule that inhibits TEAD¿YAP association and YAP-induced liver overgrowth. It is also a potent second-generation photosensitizing agent derived from porphyrin. Verteporfin is an autophagy inhibitor. Verteporfin inhibits cell proliferation and induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1119133