Learn More
Medchemexpress LLC Verteporfin | 129497-78-5 | 98.2% | 200 MG
SDP
Catalog No. 5000450904
Encompass_Preferred
Verteporfin (CL 318952) is a photosensitizer used in photodynamic therapy to eliminate abnormal blood vessels in the eye, especially those associated with age-related macular degeneration. It functions as a YAP inhibitor by disrupting YAP-TEAD interactions, inducing cell apoptosis. This compound also acts as an autophagy inhibitor, blocking autophagosome formation at an early stage.
  • Acts as a YAP inhibitor, disrupting YAP-TEAD interactions.
  • Induces cell apoptosis.
  • Inhibits autophagy by blocking autophagosome formation at an early stage.
  • Utilized as a photosensitizer for photodynamic therapy.
  • Shows light-independent anti-leukemic activity.
  • Reduces mitochondrial membrane potential in PDX cells.
  • Reverses certain drug resistances by inhibiting YAP expression.
Catalog No. 50-004-50904 Supplier Medchemexpress LLC Supplier No. HYB0146200MG
Verteporfin (CL 318952) is a photosensitizer used in photodynamic therapy to eliminate abnormal blood vessels in the eye, especially those associated with age-related macular degeneration. It functions as a YAP inhibitor by disrupting YAP-TEAD interactions, inducing cell apoptosis. This compound also acts as an autophagy inhibitor, blocking autophagosome formation at an early stage.
  • Acts as a YAP inhibitor, disrupting YAP-TEAD interactions.
  • Induces cell apoptosis.
  • Inhibits autophagy by blocking autophagosome formation at an early stage.
  • Utilized as a photosensitizer for photodynamic therapy.
  • Shows light-independent anti-leukemic activity.
  • Reduces mitochondrial membrane potential in PDX cells.
  • Reverses certain drug resistances by inhibiting YAP expression.