Learn More
Medchemexpress LLC HY-B0146 100mg , Verteporfin CL 318952 CAS:129497-78-5 Purity:98%
SDP
Catalog No. 501492760
Encompass_Preferred
Medchemexpress, HY-B0146 100mg Verteporfin CL 318952 CAS:129497-78-5 Formula:C41H42N4O8 Purity:98% Verteporfin is a benzoporphyrin derivative monoacid ring A, and can inhibit the activity of YAP . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2760 Supplier Medchemexpress LLC Supplier No. HYB0146100MG
Medchemexpress, HY-B0146 100mg Verteporfin CL 318952 CAS:129497-78-5 Formula:C41H42N4O8 Purity:98% Verteporfin is a benzoporphyrin derivative monoacid ring A, and can inhibit the activity of YAP . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.