Learn More
Medchemexpress LLC HY-B0146 50mg , Verteporfin CAS:129497-78-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB014650MG
Encompass_Preferred
HY-B0146 50mg Verteporfin CAS: 129497-78-5 Verteporfin (CL 318952) is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as age-related macular degeneration. Verteporfin is a YAP inhibitor which disrupts YAP-TEAD interactions. Verteporfin induces cell apoptosis. Verteporfinis an autophagy inhibitor that blocks autophagy at an early stage by inhibiting autophagosome formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9572