Learn More
Abcam Verteporfin (Visudyne), Benzoporphyrin derivative, 25MG
Catalog No. 502706121
Encompass_Preferred
MW 718.8 Da, Purity >96%. Benzoporphyrin derivative. Antiangiogenic agent. Produces ROS on stimulation with nonthermal red light in the presence of oxygen. Prevents corneal neovascularization and shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6121 Supplier Abcam Supplier No. AB14329425MG
MW 718.8 Da, Purity >96%. Benzoporphyrin derivative. Antiangiogenic agent. Produces ROS on stimulation with nonthermal red light in the presence of oxygen. Prevents corneal neovascularization and shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement