Learn More
JDM LAB SOLUTIONS LLC SSI Bio Vertex-LE Pipette Tips, 20 µL, Racked, Sterile, LTS-Compatible, 960/Pk.
Catalog No. NC3935292
Encompass
These 20 µL pipette tips are precision-engineered for compatibility with single- and multi-channel LiteTouch® pipettors. Manufactured from medical-grade virgin polypropylene, these racked, sterile tips incorporate proprietary resin technology to ensure low-retention performance and facilitate clean sample release.
  • Designed for light force tip attachment and ejection
  • Constructed from medical-grade virgin polypropylene
  • Rack bases and grids produced with 50% post-consumer recycled content
  • Certified free of detectable RNase, DNase, DNA, and PCR inhibitors
  • Endotoxin-free for sterile applications
Catalog No. NC3935292 Supplier JDM LAB SOLUTIONS LLC Supplier No. 41L7NS0S
These 20 µL pipette tips are precision-engineered for compatibility with single- and multi-channel LiteTouch® pipettors. Manufactured from medical-grade virgin polypropylene, these racked, sterile tips incorporate proprietary resin technology to ensure low-retention performance and facilitate clean sample release.
  • Designed for light force tip attachment and ejection
  • Constructed from medical-grade virgin polypropylene
  • Rack bases and grids produced with 50% post-consumer recycled content
  • Certified free of detectable RNase, DNase, DNA, and PCR inhibitors
  • Endotoxin-free for sterile applications