Learn More
Electron Microscopy Sciences A stainless steel rack with a fixed handle for use with large slides. Rack is grooved to take 76mm microscope slides up to 50mm wide horizontally. Accommodates up to 24 slides.
SDP
Catalog No. NC3585409
Encompass_Preferred
EMS Vertical 48 Slide Rack Only
Catalog No. NC3585409 Supplier Electron Microscopy Sciences Supplier No. 71421VR
EMS Vertical 48 Slide Rack Only