Learn More
Electron Microscopy Sciences Vertical 48 Slide Rack only
SDP
Catalog No. 50931053
Encompass_Preferred
A stainless steel rack with a fixed handle for use with large slides. Rack is grooved to take 76mm microscope slides up to 50mm wide horizontally. Accommodates up to 55 slides.
Catalog No. 50-931-053 Supplier Electron Microscopy Sciences Supplier No. 71422VR
A stainless steel rack with a fixed handle for use with large slides. Rack is grooved to take 76mm microscope slides up to 50mm wide horizontally. Accommodates up to 55 slides.