Learn More
Electron Microscopy Sciences Vertical 55-Slide Rack only
SDP
Catalog No. 50931052
Encompass_Preferred
A stainless steel rack with a fixed handle and grooves for 76 x 26 mm microscope slides held vertically for maximum use of space and solutions. Accommodates two vertical racks (#71421-VR),which totals 96 slides.
Catalog No. 50-931-052 Supplier Electron Microscopy Sciences Supplier No. 71422FR
Add to Cart
Add to Cart
A stainless steel rack with a fixed handle and grooves for 76 x 26 mm microscope slides held vertically for maximum use of space and solutions. Accommodates two vertical racks (#71421-VR),which totals 96 slides.