Learn More
Electron Microscopy Sciences Visio-Tek™ Visio-Tek Microscope Boom Stand
SDP
Catalog No. 501930537
Encompass_Preferred
Vertical/desktop rack that utilizes an adjustable sliding horizontal boom arm with up to 25 cm reach and 360° rotational capability. It is ideal for inspection applications where the ability to extend over and across a specimen is required.
Catalog No. 50-193-0537 Supplier Electron Microscopy Sciences Supplier No. 9950007
Vertical/desktop rack that utilizes an adjustable sliding horizontal boom arm with up to 25 cm reach and 360° rotational capability. It is ideal for inspection applications where the ability to extend over and across a specimen is required.