Learn More
Kinematica HMC Europe Vertical Autoclave Printers
Catalog No. 08451428 Shop All Kinematica Products
Change view
Click to view available options
Model:
HG 50/80
HV 50/80
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
08-451-428 HV 50/80
08-451-413 HG 50/80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 08-451-428 Supplier Kinematica HMC Europe Supplier No. 50003
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Print record of sterilization with an integrated printer.
Batch printer for direct recording of the sterilization data as proof of successful sterilization.
Specifications
For Use With (Application) Direct recording of sterilization data, temperature and time
Model HV 50/80