Learn More
Poltex LLC Vertical Biohazard Bag Box with Lid, 8 x 12 in. Bag (Bumpons)

Description
- Lid and back part are removable for bag removal and easy cleaning
- Rubber bumpons on base for non-slip surface
- Durable and cleanable PETG construction
- Consumables not included
- Made in the USA
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Hinged Lid, No Clasp |
| Color | Clear |
| Disposable | Reusable |
| Material | PETG |
| Sterility | Non-sterile |
| Quantity | 1 Ea. |
| Dimensions (L x W) | 4.4 x 3.6 in. |
| Length (English) | 4.4 in. |
| Width (English) | 3.6 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.