Learn More
Braintree Scientific Vertical Canister Holder, wall mountable,FC-A not included
SDP
Catalog No. 501954516
Encompass
Vertical Canister Holder - Ensures proper canister positionA canister on its side will allow waste gases to travel across the top limiting its effectiveness while a canister on a table will block the waste gases from exiting. The vertical canister holder accepts any canister thats 3 dia. or smaller. Model #FC-A is included and connects standard 1/4 tubing to the canister. The vertical holder is for passive systems only and can be tabletop or wall mountable. See model number FC-A for 90 degree adapter only. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4516 Supplier Braintree Scientific Supplier No. CHU1
Vertical Canister Holder - Ensures proper canister positionA canister on its side will allow waste gases to travel across the top limiting its effectiveness while a canister on a table will block the waste gases from exiting. The vertical canister holder accepts any canister thats 3 dia. or smaller. Model #FC-A is included and connects standard 1/4 tubing to the canister. The vertical holder is for passive systems only and can be tabletop or wall mountable. See model number FC-A for 90 degree adapter only. **Sold for research purposes only and not for human use**